missing translation for 'onlineSavingsMsg'
Learn More

1,1'-Carbonyldipyrrolidine, 98%, Thermo Scientific™

Catalog Number p-7053552
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
81759-25-3
C9H16N2O
168.24
MFCD00216641
HPTQKSXAQBHFKL-UHFFFAOYSA-N
1,1'-carbonyldipyrrolidine, di pyrrolidin-1-yl methanone, 1,1-carbonyldipyrrolidine, 1-pyrrolidine-1-carbonyl pyrrolidine, pyrrolidine, 1,1'-carbonylbis, ditetrahydro-1h-pyrrol-1-ylmethanone, carbodipiperidide, dipyrrolizinoketone, carbonyldipyrrolidine, dipyrrolidinyl ketone
736440
dipyrrolidin-1-ylmethanone
O=C(N1CCCC1)N1CCCC1
This item is not returnable. View return policy
81759-25-3
C9H16N2O
168.24
MFCD00216641
HPTQKSXAQBHFKL-UHFFFAOYSA-N
1,1'-carbonyldipyrrolidine, di pyrrolidin-1-yl methanone, 1,1-carbonyldipyrrolidine, 1-pyrrolidine-1-carbonyl pyrrolidine, pyrrolidine, 1,1'-carbonylbis, ditetrahydro-1h-pyrrol-1-ylmethanone, carbodipiperidide, dipyrrolizinoketone, carbonyldipyrrolidine, dipyrrolidinyl ketone
736440
dipyrrolidin-1-ylmethanone
O=C(N1CCCC1)N1CCCC1
Applications
Reactant in the ring-opening polymerization of six-and seven-membered cyclic pseudoureas.

Solubility
Slightly soluble in water.

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light.
TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Density 1.086 g/mL
Boiling Point 148°C to 150°C
Flash Point 113°C (235°F)
Refractive Index 1.513
Quantity 10 g
Beilstein 1637732
Sensitivity Light sensitive
Solubility Information Slightly soluble in water.
Formula Weight 168.24
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1,1'-Carbonyldipyrrolidine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'