CAS RN 7787-37-3
CAS RN 7787-37-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
297.27
Molecular Formula:
BaMoO4
InChi Key:
RCMWGBKVFBTLCW-UHFFFAOYSA-N
SMILES:
[Ba++].[O-][Mo]([O-])(=O)=O
1
–
1
of
1
results
1
Barium molybdenum oxide, 99.9% (metals basis excluding Sr), Thermo Scientific Chemicals
CAS: 7787-37-3 MDL Number: MFCD00014182 Synonym: Barium molybdate