accessibility menu, dialog, popup

UserName

CAS RN 639052-78-1

CH3ClNSHNO

CAS RN 639052-78-1

IUPAC Name: N-(5-chloro-6-methyl-1,3-benzothiazol-2-yl)-2-phenylacetamide
Synonyms: N-(5-chloro-6-methyl-1,3-benzothiazol-2-yl)-2-phenylacetamide
Molecular Weight (g/mol): 316.8
Molecular Formula: C16H13ClN2OS
InChi Key: DYHAMRNAHTWYKY-UHFFFAOYSA-N
SMILES: CC1=C(Cl)C=C2N=C(NC(=O)CC3=CC=CC=C3)SC2=C1
Color 
  • (3)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results