accessibility menu, dialog, popup

UserName

CAS RN 6385-62-2

H2OBrBrNN

CAS RN 6385-62-2

IUPAC Name: 7λ⁵,10λ⁵-diazatricyclo[8.4.0.0²,⁷]tetradeca-1(14),2,4,6,10,12-hexaene-7,10-bis(ylium) hydrate dibromide
Synonyms: diquat dication hydrate dibromide
Molecular Weight (g/mol): 362.07
Molecular Formula: C12H14Br2N2O
InChi Key: KLJOSQAQZMLLMB-UHFFFAOYSA-L
SMILES: O.[Br-].[Br-].C1C[N+]2=CC=CC=C2C2=CC=CC=[N+]12
Molecular Weight (g/mol) 
  • (1)
  • (3)
Solvent or Matrix 
  • (1)
  • (2)
Concentration 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)