CAS RN 63648-73-7
CAS RN 63648-73-7
IUPAC Name:
2-{[(benzyloxy)carbonyl]amino}pentanedioic acid
Synonyms:
2-{[(benzyloxy)carbonyl]amino}pentanedioic acid
Molecular Weight (g/mol):
281.26
Molecular Formula:
C13H15NO6
InChi Key:
PVFCXMDXBIEMQG-UHFFFAOYNA-N
SMILES:
OC(=O)CCC(NC(=O)OCC1=CC=CC=C1)C(O)=O
1
–
1
of
1
results
1
N-Benzyloxycarbonyl-D-glutamic acid, 95%
CAS: 63648-73-7 Molecular Formula: C13H15NO6 Molecular Weight (g/mol): 281.264 MDL Number: MFCD00063193 InChI Key: PVFCXMDXBIEMQG-SNVBAGLBSA-N Synonym: z-d-glu-oh,z-d-glutamic acid,n-carbobenzoxy-d-glutamic acid,z-d-glu,n-cbz-d-glutamic acid,n-benzyloxycarbonyl-d-glutamic acid,r-2-benzyloxy carbonyl amino pentanedioic acid,cbz-glu-oh,2r-2-benzyloxy carbonyl amino pentanedioic acid,cbz-d-glu PubChem CID: 1607568 IUPAC Name: (2R)-2-(phenylmethoxycarbonylamino)pentanedioic acid SMILES: C1=CC=C(C=C1)COC(=O)NC(CCC(=O)O)C(=O)O