accessibility menu, dialog, popup

UserName

CAS RN 63604-59-1

H3CHNNN

CAS RN 63604-59-1

IUPAC Name: 2-methyl-3H-imidazo[4,5-c]pyridine
Synonyms: 2-methyl-3H-imidazo[4,5-c]pyridine
Molecular Weight (g/mol): 133.15
Molecular Formula: C7H7N3
InChi Key: JWQVOMJORQXNME-UHFFFAOYSA-N
SMILES: CC1=NC2=CC=NC=C2N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results