CAS RN 634924-89-3
CAS RN 634924-89-3
IUPAC Name:
8-ethoxy-9-ethyl-9H-purin-6-amine
Synonyms:
8-ethoxy-9-ethylpurin-6-amine
Molecular Weight (g/mol):
207.24
Molecular Formula:
C9H13N5O
InChi Key:
QUGDTMONBLMLLD-UHFFFAOYSA-N
SMILES:
CCOC1=NC2=C(N)N=CN=C2N1CC