CAS RN 63484-12-8
CAS RN 63484-12-8
IUPAC Name:
methyl 5-methanesulfonyl-2-methoxybenzoate
Synonyms:
methyl 5-methanesulfonyl-2-methoxybenzoate
Molecular Weight (g/mol):
244.26
Molecular Formula:
C10H12O5S
InChi Key:
DSRPTFIZJVCPPS-UHFFFAOYSA-N
SMILES:
COC(=O)C1=CC(=CC=C1OC)S(C)(=O)=O
1
–
1
of
1
results
1
Methyl 2-Methoxy-5-(methylsulfonyl)benzoate, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.