CAS RN 63428-83-1
CAS RN 63428-83-1
IUPAC Name:
6-amino-N-{5-[(6-oxohexyl)carbamoyl]pentyl}hexanamide
Synonyms:
6-amino-N-{5-[(6-oxohexyl)carbamoyl]pentyl}hexanamide
Molecular Weight (g/mol):
341.5
Molecular Formula:
C18H35N3O3
InChi Key:
HKJOYARIOKGODS-UHFFFAOYSA-N
SMILES:
NCCCCCC(=O)NCCCCCC(=O)NCCCCCC=O