CAS RN 63283-36-3
CAS RN 63283-36-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Calcifediol Monohydrate, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
Calcifediol monohydrate, MedChemExpress
MedChemExpress Calcifediol monohydrate (25-hydroxy Vitamin D3 monohydrate), a major circulating metabolite of vitamin D3, is a potent VDR inhibitor.