accessibility menu, dialog, popup

UserName

CAS RN 630-93-3

NaOHNON

CAS RN 630-93-3

IUPAC Name: sodium 2,5-dioxo-4,4-diphenylimidazolidin-1-ide
Synonyms: sodium 2,5-dioxo-4,4-diphenylimidazolidin-1-ide
Molecular Weight (g/mol): 274.26
Molecular Formula: C15H11N2NaO2
InChi Key: FJPYVLNWWICYDW-UHFFFAOYSA-M
SMILES: [Na+].O=C1NC(C(=O)[N-]1)(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Certifications/Compliance 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results