CAS RN 630-60-4
CAS RN 630-60-4
IUPAC Name:
4-[(1R,3aS,3bR,5aS,7S,9R,9aR,9bS,10R,11aR)-3a,5a,9,10-tetrahydroxy-9a-(hydroxymethyl)-11a-methyl-7-{[(2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxy}-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]-2,5-dihydrofuran-2-one
Synonyms:
ouabain G-strophanthin strophanthin G strophanthin-G gratus strophanthin ouabaine uabaina uabanin purostrophan strodival
Molecular Weight (g/mol):
584.66
Molecular Formula:
C29H44O12
InChi Key:
LPMXVESGRSUGHW-HBYQJFLCSA-N
SMILES:
C[C@@H]1O[C@@H](O[C@H]2C[C@@H](O)[C@]3(CO)[C@H]4[C@H](O)C[C@]5(C)[C@H](CC[C@]5(O)[C@@H]4CC[C@]3(O)C2)C2=CC(=O)OC2)[C@H](O)[C@H](O)[C@H]1O