accessibility menu, dialog, popup

UserName

CAS RN 630-56-8

H3COO(R)(S)(R)O(R)CH3(S)(S)CH3CH3O

CAS RN 630-56-8

IUPAC Name: (1R,3aS,3bR,9aR,9bS,11aS)-1-acetyl-9a,11a-dimethyl-7-oxo-1H,2H,3H,3aH,3bH,4H,5H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl hexanoate
Synonyms: proge hydroxyprogesterone caproate proluton depot lutate primolut depot hydroxyprogesterone hexanoate hylutin luteocrin hormofort oxyprogesterone caproate
Molecular Weight (g/mol): 428.61
Molecular Formula: C27H40O4
InChi Key: DOMWKUIIPQCAJU-LJHIYBGHSA-N
SMILES: CCCCCC(=O)O[C@@]1(CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C)C(C)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results