CAS RN 630-07-9
CAS RN 630-07-9
IUPAC Name:
pentatriacontane
Synonyms:
N-pentatriacontane
Molecular Weight (g/mol):
492.96
Molecular Formula:
C35H72
InChi Key:
VHQQPFLOGSTQPC-UHFFFAOYSA-N
SMILES:
CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC