accessibility menu, dialog, popup

UserName

CAS RN 6282-02-6

HOHNO

CAS RN 6282-02-6

IUPAC Name: N-(hydroxymethyl)benzamide
Synonyms: benzamide, N-(hydroxymethyl)-
Molecular Weight (g/mol): 151.17
Molecular Formula: C8H9NO2
InChi Key: UOUBPDZUBVJZOQ-UHFFFAOYSA-N
SMILES: OCNC(=O)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results