accessibility menu, dialog, popup

UserName

CAS RN 62780-89-6

ClNOHN

CAS RN 62780-89-6

IUPAC Name: 1-(3-chloropropyl)-2,3-dihydro-1H-1,3-benzodiazol-2-one
Synonyms: 1-(3-chloropropyl)-3H-1,3-benzodiazol-2-one
Molecular Weight (g/mol): 210.66
Molecular Formula: C10H11ClN2O
InChi Key: GUMPYDGUYXOYML-UHFFFAOYSA-N
SMILES: ClCCCN1C(=O)NC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results