CAS RN 62758-15-0
CAS RN 62758-15-0
IUPAC Name:
disodium 4-{2-[4-(dimethylamino)phenyl]diazen-1-yl}benzene-1-sulfonate 4-{[4-(ethylamino)-3-methylphenyl][(1E)-4-(ethylimino)-3-methylcyclohexa-2,5-dien-1-ylidene]methyl}-3-sulfobenzene-1-sulfonate
Synonyms:
disodium methyl orange 4-{[4-(ethylamino)-3-methylphenyl][(1E)-4-(ethylimino)-3-methylcyclohexa-2,5-dien-1-ylidene]methyl}-3-sulfobenzenesulfonate
Molecular Weight (g/mol):
865.94
Molecular Formula:
C39H41N5Na2O9S3
InChi Key:
QZATZDKHJDOZFD-DGVIJJPGSA-L
SMILES:
[Na+].[Na+].CN(C)C1=CC=C(C=C1)N=NC1=CC=C(C=C1)S([O-])(=O)=O.CCNC1=CC=C(C=C1C)C(=C1\C=CC(=NCC)C(C)=C1)\C1=CC=C(C=C1S(O)(=O)=O)S([O-])(=O)=O
1
–
1
of
1
results
1
Promotions Available
Thermo Scientific Chemicals Methyl Orange-xylene cyanol solution in water, pure