CAS RN 62625-31-4
CAS RN 62625-31-4
IUPAC Name:
sodium 4-[3-(4-hydroxy-2-methylphenyl)-1,1-dioxo-3H-2,1λ⁶-benzoxathiol-3-yl]-3-methylbenzen-1-olate
Synonyms:
sodium 4-[3-(4-hydroxy-2-methylphenyl)-1,1-dioxo-2,1λ⁶-benzoxathiol-3-yl]-3-methylbenzenolate
Molecular Weight (g/mol):
404.41
Molecular Formula:
C21H17NaO5S
InChi Key:
ALWHNCXHKCVHCK-UHFFFAOYNA-M
SMILES:
[Na+].CC1=CC(O)=CC=C1C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C([O-])C=C1C
1
–
3
of
3
results
1
m-Cresol Purple sodium salt
CAS: 62625-31-4 Molecular Formula: C21H17NaO5S MDL Number: MFCD00010177 Synonym: m-Cresol Purple, water soluble; m-Cresolsulfonephthalein sodium salt
2
m-Cresol Purple, sodium salt, pure, water soluble
CAS: 62625-31-4 Molecular Formula: C21H17NaO5S MDL Number: MFCD00010177 Synonym: m-Cresol Purple, sodium salt
3
m-Cresol Purple sodium salt, 0.04% w/v aq. soln., Thermo Scientific™
CAS: 62625-31-4 MDL Number: MFCD00005871