CAS RN 62351-47-7
CAS RN 62351-47-7
IUPAC Name:
magnesium(2+) 3,5-difluorobenzen-1-ide bromide
Synonyms:
magnesium(2+) 3,5-difluorobenzen-1-ide bromide
Molecular Weight (g/mol):
217.3
Molecular Formula:
C6H3BrF2Mg
InChi Key:
XQNUHMQSOMLVGM-UHFFFAOYSA-M
SMILES:
[Mg++].[Br-].FC1=CC(F)=C[C-]=C1