CAS RN 623-68-7
CAS RN 623-68-7
IUPAC Name:
(2E)-but-2-enoyl (2E)-but-2-enoate
Synonyms:
2-butenoic acid, anhydride
Molecular Weight (g/mol):
154.17
Molecular Formula:
C8H10O3
InChi Key:
VJDDQSBNUHLBTD-GGWOSOGESA-N
SMILES:
C\C=C\C(=O)OC(=O)\C=C\C