accessibility menu, dialog, popup

UserName

CAS RN 61951-86-8

NaNaH3CNONHNNHNClNClNSOOOSOOOH2NOOCH3(Z)

CAS RN 61951-86-8

IUPAC Name: disodium 4-{2-[(3Z)-5-carbamoyl-1-ethyl-4-methyl-2,6-dioxo-1,2,3,6-tetrahydropyridin-3-ylidene]hydrazin-1-yl}-6-[(4,6-dichloro-1,3,5-triazin-2-yl)amino]benzene-1,3-disulfonate
Synonyms: disodium 4-{2-[(3Z)-5-carbamoyl-1-ethyl-4-methyl-2,6-dioxopyridin-3-ylidene]hydrazin-1-yl}-6-[(4,6-dichloro-1,3,5-triazin-2-yl)amino]benzene-1,3-disulfonate
Molecular Weight (g/mol): 667.36
Molecular Formula: C18H14Cl2N8Na2O9S2
InChi Key: JUNRGNKPIKIXLL-BGBYJAETSA-L
SMILES: [Na+].[Na+].CCN1C(=O)\C(=N/NC2=CC(NC3=NC(Cl)=NC(Cl)=N3)=C(C=C2S([O-])(=O)=O)S([O-])(=O)=O)C(C)=C(C(N)=O)C1=O
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results