CAS RN 616-04-6
CAS RN 616-04-6
IUPAC Name:
1-methylimidazolidine-2,4-dione
Synonyms:
1-methylhydantoin dioxy-creatinine
Molecular Weight (g/mol):
114.1
Molecular Formula:
C4H6N2O2
InChi Key:
RHYBFKMFHLPQPH-UHFFFAOYSA-N
SMILES:
CN1CC(=O)NC1=O
1
–
1
of
1
results
1
N-Methylhydantoin, MedChemExpress
MedChemExpress N-Methylhydantoin is a product of degradation of creatinine by bacteria.