CAS RN 6147-11-1
CAS RN 6147-11-1
IUPAC Name:
1,3,6-trihydroxy-7-methoxy-2,8-bis(3-methylbut-2-en-1-yl)-9H-xanthen-9-one
Synonyms:
mangostin α-mangostin
Molecular Weight (g/mol):
410.47
Molecular Formula:
C24H26O6
InChi Key:
GNRIZKKCNOBBMO-UHFFFAOYSA-N
SMILES:
COC1=C(O)C=C2OC3=CC(O)=C(CC=C(C)C)C(O)=C3C(=O)C2=C1CC=C(C)C
1
–
1
of
1
results
1
alpha-Mangostin, MedChemExpress
MedChemExpress alpha-Mangostin (α-Mangostin) is a dietary xanthone with broad biological activities, such as antioxidant, anti-allergic, antiviral, antibacterial, anti-inflammatory and anticancer effects. It is an inhibitor of mutant IDH1 (IDH1-R132H) with a Ki of 2.85 μM.