CAS RN 6142-90-1
CAS RN 6142-90-1
IUPAC Name:
furan
Synonyms:
furan divinylene oxide tetrole furfuran oxacyclopentadiene rcra waste number U124 axole hsdb 89
Molecular Weight (g/mol):
68.08
Molecular Formula:
C4H4O
InChi Key:
YLQBMQCUIZJEEH-UHFFFAOYSA-N
SMILES:
O1C=CC=C1
1
–
2
of
2
results
1
Furan-d4, CDN
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
2
Furan D4 1000 μg/mL in Methanol, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality