CAS RN 6139-83-9
CAS RN 6139-83-9
IUPAC Name:
4-chloro-1,1-diethoxybutane
Synonyms:
4-chloro-1,1-diethoxybutane
Molecular Weight (g/mol):
180.67
Molecular Formula:
C8H17ClO2
InChi Key:
JGGRHRMHOUWCDX-UHFFFAOYSA-N
SMILES:
CCOC(CCCCl)OCC
1
–
1
of
1
results
1
4-Chloro-1,1-diethoxybutane (4-Chlorobutyraldehyde Diethyl Acetal), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.