CAS RN 61368-63-6
CAS RN 61368-63-6
IUPAC Name:
9-[4'-({[hydroxy({[hydroxy(phosphonatooxy)phosphoryl]oxy})phosphoryl]oxy}methyl)-2,4,6-trinitrospiro[cyclohexane-1,2'-furo[3,4-d][1,3]dioxole]-2,4-dien-6'-yl]-6,9-dihydro-5H-purin-6-iminium
Synonyms:
9-[4'-({[hydroxy([hydroxy(phosphonatooxy)phosphoryl]oxy)phosphoryl]oxy}methyl)-2,4,6-trinitrospiro[cyclohexane-1,2'-furo[3,4-d][1,3]dioxole]-2,4-dien-6'-yl]-5H-purin-6-iminium
Molecular Weight (g/mol):
713.23
Molecular Formula:
C16H12N8O19P3
InChi Key:
SWLXZTUOMCLGJI-UHFFFAOYNA-M
SMILES:
OP(=O)(OCC1=C2OC3(OC2=C(O1)N1C=NC2C1=NC=NC2=[NH2+])C(C=C(C=C3[N+]([O-])=O)[N+]([O-])=O)[N+]([O-])=O)OP(O)(=O)OP([O-])([O-])=O