accessibility menu, dialog, popup

UserName

CAS RN 2384-85-2

H3CCH3

CAS RN 2384-85-2

IUPAC Name: dec-3-yne
Synonyms: 3-decyne
Molecular Weight (g/mol): 138.25
Molecular Formula: C10H18
InChi Key: JUWXVJKQNKKRLD-UHFFFAOYSA-N
SMILES: CCCCCCC#CCC
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results