accessibility menu, dialog, popup

UserName

CAS RN 143983-91-9

InClClCl

CAS RN 143983-91-9

IUPAC Name: trichloroindigane hydrate
Synonyms: indium(III) chloride hydrate
Molecular Weight (g/mol): 239.18
Molecular Formula: Cl3H2InO
InChi Key: KYCHGXYBBUEKJK-UHFFFAOYSA-K
SMILES: O.Cl[In](Cl)Cl
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results