accessibility menu, dialog, popup

UserName

CAS RN 14284-95-8

OOOOOOCCCH3CH3CCH3H3CCH3H3CHHHTb

CAS RN 14284-95-8

IUPAC Name: tris(4-hydroxypent-3-en-2-one) terbium
Synonyms: tris(4-hydroxypent-3-en-2-one) terbium
Molecular Weight (g/mol): 459.28
Molecular Formula: C15H24O6Tb
InChi Key: HEJGWJNGLVOYLY-UHFFFAOYSA-N
SMILES: [Tb].CC(O)=CC(C)=O.CC(O)=CC(C)=O.CC(O)=CC(C)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results