accessibility menu, dialog, popup

UserName

CAS RN 109545-72-4

OOI

CAS RN 109545-72-4

IUPAC Name: (2-carboxylatophenyl)(phenyl)iodanium
Synonyms: (2-carboxylatophenyl)(phenyl)iodanium
Molecular Weight (g/mol): 324.12
Molecular Formula: C13H9IO2
InChi Key: LUGVQQXOGHCZNN-UHFFFAOYSA-N
SMILES: [O-]C(=O)C1=CC=CC=C1[I+]C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results