Learn More
Fisherbrand™ Zinc Alloy Bossheads
For securely keeping clamps in place on laboratory scaffolding. Fisherbrand™ Zinc Alloy Bossheads come with plated clamping screws with plastic heads.
Brand:  Fisherbrand™
Additional Details : Weight : 0.45000kg
Product Code. 10117184
  • 363.00 NOK / Pack of 5
Estimated Shipment: 24-05-2024
Description
For securely keeping clamps in place on laboratory scaffolding. Fisherbrand™ Zinc Alloy Bossheads come with plated clamping screws with plastic heads.
  • For rods up to 16mm in diameter
  • Supplied in packs of 5
Specifications
Specifications
5/Pk.
Zinc Alloy
For use with rod up to 16mm diameter
Bosshead Clamp
Plastic headed clamping screws