Learn More
Cytiva Whatman™ GV 025 Series Glass Vacuum Filtration Manifolds, Replacement Parts
Replacement Parts for Whatman™ GV 025 Series Vacuum Filtration Manifolds. Includes flasks, supports, funnels, frits, centering rings, and clamps.
Brand:  Cytiva 10441001
959.50 NOK valid until 2024-12-31
Use promo code "23266" to get your promotional price.
Additional Details : Weight : 0.13000kg
 View more versions of this product
Product Code. 11354914
  • 1010.00 NOK / Each
Estimated Shipment: 05-06-2024
Description
  • Description: Replacement Part for GV 025 Series Glass Vacuum Filtration Manifolds
  • For Use With (Application): Vacuum Filtration
Specifications
Specifications
Replacement Part for GV 025 Series Glass Vacuum Filtration Manifold
Borosilicate Glass
GV 025/0/01
GV025
60 mL
Manifold Model GV 025/0
Vacuum Filtration
1 Ea.
Funnel