Learn More
Fisherbrand™ Triangular PTFE Stir Bars
Particularly effective for dissolving solids and mixing sediments due to scraping-like action on the base of the container. The triangular PTFE stir bar provides stable stirring action for flat-bottom containers.
Brand:  Fisherbrand™
Additional Details : Weight : 0.04500kg
 View more versions of this product
Product Code. 11838872
  • 269.00 NOK / Pack of 10
Estimated Shipment: 17-06-2024
Description
  • Stable, general-purpose magnetic stir bar
  • Particularly effective for dissolving solids and mixing sediments due to scraping-like action on the base of the container
  • Autoclavable
Specifications
Specifications
PTFE encapsulated Alnico V magnet core
9mm
9mm
Containers & Vessels / Stirrer Hot Plates
Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
Triangle
35 x 9mm
White
5/Pk.
Yes
35 mm