Learn More
asecos™ Stainless Steel Storage Can with Dosing Tap
The Stainless Steel Safety Canister is ideal for the safe distribution and dosing of flammable liquids.
1934.70 NOK - 2469.29 NOK
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
17876516
|
asecos™
40084 |
1 L |
1934.70 NOK
Each |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
17866476
|
asecos™
40085 |
2 L |
2011.07 NOK
Each |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
17886586
|
asecos™
40086 |
5 L |
2469.29 NOK
Each |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
Description
- With fine dosing tap made of stainless steel (1.4301), incl. vent, PTFE seal, integrated flame barrier (removable) and plastic protective ring
- Incl. multilingual safety sticker to designate the contents
Specifications
Gray | |
Stainless Steel |