Learn More
Lenz Laborglasinstrumente™ Stainless Steel Fork Clamps
Brand: Lenz Laborglasinstrumente™ 01103514
Additional Details : Weight : 0.01600kg
Specifications
10/Pk. | |
Stainless Steel | |
Conical Ground Joint: NS 14/23 |
Fork Clamp | |
For conical ground joints |