Learn More
Fisherbrand™ Septum for 10-425 screw thread cap
Most septa are laminated on one side with PTFE for high chemical resistance and an inert barrier between sample and carrier material.
392.00 NOK - 4310.00 NOK
Product Code | Brand | Color | Material | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Material | Price | Quantity & Availability | ||||
11350850
|
Fisherbrand™
|
Red, White | PTFE, Silicone |
2290.00 NOK
Pack of 1000 |
Estimated Shipment: 04-06-2024
Log in to see stock. |
||||
11844820
|
Fisherbrand™
|
Blue, White | PTFE, Silicone |
4310.00 NOK
Pack of 1000 |
In Stock
Log in to see stock. |
||||
11824820
|
Fisherbrand™
|
Red-Orange, Transparent | Natural Rubber, TEF |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10422614
|
Fisherbrand™
|
White | PTFE Virginal |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
11834820
|
Fisherbrand™
|
Red, White | PTFE, Silicone |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- Septa