Learn More
R&L Enterprises™ Bright Stainless Steel Rods
An excellent option for the construction of retort stands in the laboratory. R&L Enterprises™ Bright Stainless Steel Rods are durable and corrosion resistant.
Marca: R&L Enterprises™ 3109/11
Dettagli aggiuntivi : Weight : 0.44280kg
Descrizione
- Durable
- Easy-to-clean
- Corrosion resistant
- Stainless steel
Specifica
Stainless Steel | |
12.5 mm | |
Plain (without thread) |
500 mm | |
Rod | |
For V Type Stand |