Learn More
Hettich Lab Technology™ Reductor
Hettich Lab Technology™ Reductor is designed to accommodate customized centrifuge workloads.
436.00 NOK - 1575.00 NOK
Product Code | Brand | Capacity | For Use With (Equipment) | Max. Tube Diameter | Max. Tube Length | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity | For Use With (Equipment) | Max. Tube Diameter | Max. Tube Length | Price | Quantity & Availability | ||
10757394
|
Hettich Lab Technology™
1631 |
15 mL | Rotor 1137, 1016 | 17 mm | 120 mm |
461.00 NOK
Each |
Estimated Shipment: 09-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
10685493
|
Hettich Lab Technology™
1633 |
25 mL | Rotor 1137, 1016 | 24 mm | 100 mm |
461.00 NOK
Each |
Estimated Shipment: 09-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
10592682
|
Hettich Lab Technology™
1446 |
50 mL | Rotor 1620A, 1720, 4794, 1446, 4246 | 29 mm | 107 mm |
436.00 NOK
Each |
Estimated Shipment: 09-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
10392782
|
Hettich Lab Technology™
1451 |
15 mL | Rotor 1620A, 1720, 1792, 4794, 5615, 4246 | 17 mm | 100 mm |
695.00 NOK
Each |
Estimated Shipment: 09-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
10703704
|
Hettich Lab Technology™
1635 |
9 to 10, 10, 15 mL | Rotor 1016, 1137 | 17 mm | 100 mm |
461.00 NOK
Each |
Estimated Shipment: 09-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
10711553
|
Hettich Lab Technology™
1466 |
15 mL | Rotor 1620A, 1720, 4794, 5615, 4246 | 17 mm | 120 mm |
436.00 NOK
Each |
Estimated Shipment: 09-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
12013884
|
Hettich Lab Technology™
1632 |
7 mL | Rotor 1137, 1016 | 12 mm | 100 mm |
461.00 NOK
Each |
Estimated Shipment: 09-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
14871482
|
Hettich Lab Technology™
1777 |
85/94 mL | Carrier 1752 | 38 mm | 106 mm |
1575.00 NOK
Each |
Estimated Shipment: 19-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
Description
- Available in a variety of configurations
- Configurations capable of accommodating a host of customized centrifuge workloads
Specifications
Reductor | |
1 Pack |