Learn More
Fisherbrand™ Polycarbonate Membranes
Fisherbrand™ Polycarbonate Membranes smooth flat surface and hexibit very low extractables levels.
Brand: Fisherbrand™
Additional Details : Weight : 0.01000kg
Description
- Track-etched PC membranes manufactured from high-quality polycarbonate film
Specifications
White | |
0.2 μm | |
10mL/min./cm2 | |
3L/min./cm2 | |
0.25 mm | |
Yes |
Polycarbonate | |
Non-sterile | |
Fluids clarification, environnemental analysis, cytology, parasitology, bioassays, epifluorescence microscopy, analysis of biological cells and particles | |
100 Pack | |
Plain Disc | |
47 mm |