Learn More
Quickfit™ 27BU Organic Chemistry Kit
Ideal organic chemistry kit for use in schools, colleges, and universities. Quickfit™ 27BU Organic Chemistry Kit is perfect for introducing students to the principal techniques of organic chemistry.
Brand: Quickfit™ 27BU
3686.00 NOK valid until 2024-12-31
Use promo code "23259" to get your promotional price.
Additional Details : Weight : 0.60000kg
Description
- The most popular Quickfit™ set for schools, colleges and universities
- Comprises nine basic items of Quickfit™ glassware all with size 14/23 ground glass joints for compatibility
- Allows the construction of at least 8 assemblies including steam distillation, reflux with addition and preparation
- Scale based on starting material quantities up to 30g
- Housed in a robust cardboard box with foam rubber liner to provide safe storage and transit
- Supplied with a CD-ROM which includes practical details of some typical assemblies
Specifications
Organic Chemistry Kit | |
Pear shaped flask, Stillhead, Liebig condenser, Screwcap adapter, Receiver adapter, Air leak/steam inlet tube, Dropping funnel with GP Rotoflo tap, Stopper, Thermometer |