Learn More
Thermo Scientific™ Megafuge ST4F Plus, 230V TX-1000 Tissue Culture Package
Waste no time finding the right part numbers for your application! The application driven centrifuge package ensures convenient, fast ordering of all components necessary to run a specific protocol.
Brand:  Thermo Scientific™ 75017961
Additional Details : Weight : 183.00000kg
Product Code. 17102489
  • 199225.00 NOK / 1 set
Estimated Shipment: 20-09-2024
Description
Waste no time finding the right part numbers for your application! The application driven centrifuge package ensures convenient, fast ordering of all components necessary to run a specific protocol. Exceptional capacity to process 40 x 50 mL conicals in one run, under biocontainment certified lids. Contains centrifuge, TX-1000 rotor, buckets, biocontainment lids, 50 mL conical adapters (maximum capacity 40 x 50 mL), 15 mL conical adapter (maximum capacity 96 x 15 mL).
  • Easy to Use:
    • Intuitive controls simplify operation and provide detailed information on a wide range of operating parameters and processing status for optimal performance.
    • Modern industrial design supports continuous improvement in rotor loading ergonomics, user safety and current needed certifications
    • Motorized lid latch enables one finger downward motion for easiest closing and locking of centrifuge lid, even on tall benches.
    Thermo Scientific™ Auto-Lock™ III Rotor System:
    • Secure locking system allows easy push-button installation and exchange of rotors.
    • System saves time when switching between applications.
    • Easy access to rotor chamber offers quick cleaning, a healthier working environment and longer unit life.
    Thermo Scientific™ ClickSeal™ Bucket Sealing System:
    • Biocontainment solution seals with a snap; eliminates screw caps and complicated clips.
    • System has glove-friendly, one-handed open/close capability.
    • System is certified by CAMR™ in Porton Down, UK.
    Thermo Scientific™ SMARTSpin™ Technology:
    • Advanced rotor management system maximizes acceleration, braking and residual load imbalance for each rotor and bucket option:
    • Technology optimizes safety and improves separations.
    Space-Saving Design:
    • Compact dimensions optimize work area and ergonomic height and curved design ensure easy sample loading and unloading and easy cleaning.
    Energy Efficient:
    • System offers up to 40% energy consumption savings on industry standard protocols, such as standard blood separation or conical tubes processing.
Specifications
Specifications
Direct, Brushless induction low profile motor with choices of 9 acceleration, 10 deceleration rates
Part Numbers In Pk. 75009961 (centrifuge),75003017(rotor),75003001(set of buckets),75007309(lids),75007306(set of adapters),75003674(set of adapters)
General Purpose Centrifuge Series