missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ChargeSwitch™ Direct gDNA Purification Kit, 96-well

ChargeSwitch Direct 96 gDNA Kit is designed for rapid and efficient isolation of gDNA from fresh or frozen samples.

Brand:  Invitrogen™ CS11205

Additional Details : Weight : 0.44000kg

Product Code. 10405042

  • 1866.00 NOK / Each
Estimated Shipment: 13-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Includes: ChargeSwitch Plate Binding and lysis Buffer, ChargeSwitch Wash Buffers, ChargeSwitch Elution Buffer, ChargeSwitch Direct 96 gDNA Plates, and Plate Seals.

Description

Description

  • ChargeSwitch Direct 96 gDNA Kit is designed for rapid and efficient isolation of gDNA from fresh or frozen samples
  • Utilizes 96-well plates coated with ChargeSwitch nucleic acid-binding surface, whose ionic charge can be switched by changing pH of surrounding buffer
  • At low pH, positive charge of ChargeSwitch surface binds negatively charged nucleic acids
  • Proteins and other contaminants that do not bind are washed away, leaving pure DNA ready for downstream applications
  • Successful yields from very small samples volumes up to 10 μL blood or 2 x 104cells
  • Highly pure DNA isolated using gentle buffers—no guanidine or ethanol
  • No sample transfer that can lead to error or contamination—direct amplification in the same well as DNA binding without elution
  • Adaptable to your throughput needs—compatible with most automated Liquid handling platforms

Order Info

Shipping Condition: Room Temperature

Specifications

Specifications

Store at room temperature.
ChargeSwitch Chemistry
PCR, Southern Blotting, Sequencing, Cloning
High-throughput Compatible
< 100 μL Blood Samples (Small-scale)
Direct gDNA Purification Kit
Room Temperature
Kit
Blood, Cells
Fewer resources used and less waste, less hazardous
96 Preps
ChargeSwitch™
96 Preps
Genomic DNA
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.