Learn More
Fisherbrand™ Grade 911 Universal pH Indicator Paper
Provides rapid and precise readings for measuring pH.
Brand:  Fisherbrand™
Additional Details : Weight : 0.01530kg
Product Code. 11773554
  • 56.30 NOK / Each
Estimated Shipment: 27-05-2024
Description
  • Filter paper soaked with one or several colored indicators of pH
  • Dip indicator paper into the liquid to be tested for a few seconds and remove it from the test solution and compare the resultant colors with the color scale printed on the cover
  • Provided on a dispenser roll of 5m
  • pH range of 1 to 11
Specifications
Specifications
1 to 11
Measuring pH
5 m Dispenser Roll
Indicator strips