Learn More
Cole-Parmer™ Stuart™ SW6 Flocculator
Designed for repeatable conditions between samples and from run to run
Brand:  Cole-Parmer™ 99551-05
Additional Details : Weight : 9.21600kg
Product Code. 11436568
  • 32370.00 NOK / Each
Estimated Shipment: 24-06-2024
Description
  • Accepts six samples.
  • For flocculation (jar) tests on water and effluent samples.
  • Designed for repeatable conditions between samples and from run to run.
  • A digital display clearly indicates the speed of the rotational stainless steel paddles, which can be varied from 25 to 250rpm.
  • The digital timer can be set to count down from 1 to 99 minutes. After count down, the paddles stop and an alert sounds. The timer then counts up in minutes in order to measure settling time.
  • Two pre-set programs allow routine speed and time parameters to be input and then selected by simply pressing one button. Parameters are stored in the memory and can be recalled or altered at any time.
  • Intuitive touch sensitive keypad is totally flat for easy cleaning.
  • Sample monitoring is assisted by diffused base illumination and black background.
  • A simultaneous coagulant doser is available as an accessory.
Specifications
Specifications
25 to 250 rpm
750 x 210 x 460 mm
Touch-sensitive keypad, digital display
Flocculator
6
230V, 50Hz, 200W
0 to 99 min.
17 kg