Learn More
3M™ Aura™ FFP1-FFP3 Particulate Respirators
3M™ Aura™ Particulate Respirator provides breathing protection with a flat-fold, 3-panel design, low breathing resistance and an embossed, sculpted top panel that can help reduce eyewear fogging.
131.74 NOK - 34035.00 NOK
Product Code | Brand | Filter Type | Model | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Filter Type | Model | Price | Quantity & Availability | ||||
10583051
|
3M™
7000088723 |
FFP2 | 9320 |
575.00 NOK
Pack of 20 |
Estimated Shipment: 18-04-2024
Log in to see stock. |
||||
View alternative products » | |||||||||
10116544
|
3M™
7000088722 |
FFP2 | 9322+ |
495.00 NOK
Pack of 10 |
Estimated Shipment: 22-04-2024
Log in to see stock. |
||||
View alternative products » | |||||||||
11787574
|
3M™
7000062383 |
FFP2 | PF9320 |
172.00 NOK
Pack of 5 |
Estimated Shipment: 16-05-2024
Log in to see stock. |
||||
View alternative products » | |||||||||
11707584
|
3M™
7000062384 |
FFP3 | PF9332 |
405.90 NOK
Pack of 5 Qty Price Break Available |
In Stock
Log in to see stock. |
||||
View alternative products » | |||||||||
16523771
|
3M™
7000089312 |
FFP3 | 9332+ P3 valved |
24425.00 NOK
Pack of 330 |
Estimated Shipment: 22-04-2024
Log in to see stock. |
||||
16684912
|
3M™
7000088730 |
FFP2 | 9322+ P2 valved |
12565.00 NOK
Pack of 330 |
Estimated Shipment: 24-05-2024
Log in to see stock. |
||||
16674902
|
3M™
7100231525 |
FFP3 | 9330+ P3 |
34035.00 NOK
Pack of 440 |
Estimated Shipment: 16-05-2024
Log in to see stock. |
||||
11777574
|
3M™
7000062381 |
FFP1 | PF9312 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
View alternative products » | ||||
11717574
|
3M™
7000088725 |
FFP1 | 9310 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
View alternative products » | ||||
11797574
|
3M™
7000062382 |
FFP2 | PF9322 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
View alternative products » | ||||
17237620
|
3M™
7000088724 |
FFP1 | 9312 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
View alternative products » | ||||
12672545
|
3M™
7000088724 |
FFP1 | 9312 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
View alternative products » | ||||
17257620
|
3M™
7000088812 |
FFP3 | 9330 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
View alternative products » | ||||
12602555
|
3M™
7000088812 |
FFP3 | 9330 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
View alternative products » | ||||
Description
Providing a unique three-panel design for greater comfort and easy communication. 3M™ Aura™ 9300 Series Particulate Respirators cover a range of applications requiring FFP1, FFP2 and FFP3 protection.
- Individually packaged to avoid contamination
- Soft cover web on inner panel ensures greater comfort
- Lightweight and foldable
- Evenly tensioned headbands to relieve strap pressure
- Centrally positioned cool flow valve reduces heat build-up and makes breathing easier and cooler