Learn More
SCA Hygiene Products™ Facial Tissue
SCA Hygiene Products™ Facial Tissue: 2-ply tissues (100 tissues/box).
Brand:  SCA Hygiene Products™ 140280
820.38 NOK valid until 2024-05-31
Use promo code "21679" to get your promotional price.
Additional Details : Weight : 4.56300kg
Product Code. 13599450
  • 925.00 NOK / Pack of 3000
Estimated Shipment: 14-06-2024
Description
  • 100 tissues per box
  • 2-ply
Specifications
Specifications
Facial Tissues
200 mm
Can be Used in F1 Facial Dispensers
208 mm
White
2-ply