Learn More
Lenz Laborglasinstrumente™ Duran Glass Centrifuge Tubes
Constructed from borosilicate glass. Lenz Laborglasinstrumente™ Duran Glass Centrifuges Tubes are used to separate solids from liquids in chemical solutions.
Brand:  Lenz Laborglasinstrumente™ B0456511
Additional Details : Weight : 1.19700kg
Product Code. 10548840
  • 22.00 NOK / Each
Estimated Shipment: 22-05-2024
Description
  • Borosilicate glass
  • Round bottom
Specifications
Specifications
12 mL
1
100 mm
Borosilicate Glass
Round
Round