Learn More
DWK Life Sciences DURAN™ Filter Funnel
Filter funnels are filters that are used for solid-liquid separation by means of vacuum.
450.00 NOK - 7230.00 NOK
Specifications
Material | Boro 3.3 |
---|---|
Autoclavable | Yes |
For Use With (Application) | Qualitative inorganic analysis and preparative chemistry; Fine filtration; Fine filtration; Filtration at reduced pressure; Filtering |
Quantity | 1 Ea. |
Product Code | Brand | Capacity (Metric) | Porosity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Porosity | Price | Quantity & Availability | ||||
10241701
|
DWK Life Sciences
258521106 |
75 mL | 1 |
535.00 NOK
Each |
Estimated Shipment: 21-05-2024
Log in to see stock. |
||||
10517911
|
DWK Life Sciences
258521209 |
75 mL | 2 |
535.00 NOK
Each |
Estimated Shipment: 04-06-2024
Log in to see stock. |
||||
10684931
|
DWK Life Sciences
258521303 |
75 mL | 3 |
510.00 NOK
Each |
Estimated Shipment: 24-05-2024
Log in to see stock. |
||||
10778511
|
DWK Life Sciences
258521406 |
75 mL | 4 |
555.00 NOK
Each |
Estimated Shipment: 05-06-2024
Log in to see stock. |
||||
10048060
|
DWK Life Sciences
258522102 |
125 mL | 1 |
725.00 NOK
Each |
Estimated Shipment: 21-05-2024
Log in to see stock. |
||||
10568481
|
DWK Life Sciences
258522308 |
125 mL | 3 |
705.00 NOK
Each |
Estimated Shipment: 04-06-2024
Log in to see stock. |
||||
10242131
|
DWK Life Sciences
258522402 |
125 mL | 4 |
800.00 NOK
Each |
Estimated Shipment: 04-06-2024
Log in to see stock. |
||||
10508291
|
DWK Life Sciences
258526301 |
4000 mL | 3 |
6195.00 NOK
Each |
Estimated Shipment: 21-05-2024
Log in to see stock. |
||||
10706981
|
DWK Life Sciences
258520101 |
50 mL | 1 |
473.00 NOK
Each |
Estimated Shipment: 21-05-2024
Log in to see stock. |
||||
10028000
|
DWK Life Sciences
258520204 |
50 mL | 2 |
473.00 NOK
Each |
Estimated Shipment: 21-05-2024
Log in to see stock. |
||||
10747171
|
DWK Life Sciences
258520307 |
50 mL | 3 |
450.00 NOK
Each |
In Stock
Log in to see stock. |
||||
10424461
|
DWK Life Sciences
258520401 |
50 mL | 4 |
478.00 NOK
Each |
Estimated Shipment: 19-06-2024
Log in to see stock. |
||||
10304021
|
DWK Life Sciences
258520504 |
50 mL | 5 |
865.00 NOK
Each |
Estimated Shipment: 21-05-2024
Log in to see stock. |
||||
10675311
|
DWK Life Sciences
258521509 |
75 mL | 5 |
955.00 NOK
Each |
Estimated Shipment: 21-05-2024
Log in to see stock. |
||||
10303401
|
DWK Life Sciences
258522205 |
125 mL | 2 |
725.00 NOK
Each |
Estimated Shipment: 21-05-2024
Log in to see stock. |
||||
10242181
|
DWK Life Sciences
258522505 |
125 mL | 5 |
1430.00 NOK
Each |
Estimated Shipment: 04-06-2024
Log in to see stock. |
||||
10403331
|
DWK Life Sciences
258523107 |
500 mL | 1 |
1175.00 NOK
Each |
Estimated Shipment: 21-05-2024
Log in to see stock. |
||||
10057940
|
DWK Life Sciences
258523201 |
500 mL | 2 |
1175.00 NOK
Each |
Estimated Shipment: 21-05-2024
Log in to see stock. |
||||
10313011
|
DWK Life Sciences
258523304 |
500 mL | 3 |
1175.00 NOK
Each |
Estimated Shipment: 04-06-2024
Log in to see stock. |
||||
10570202
|
DWK Life Sciences
258523407 |
500 mL | 4 |
1405.00 NOK
Each |
Estimated Shipment: 04-06-2024
Log in to see stock. |
||||
10494451
|
DWK Life Sciences
258523501 |
500 mL | 5 |
2955.00 NOK
Each |
Estimated Shipment: 21-05-2024
Log in to see stock. |
||||
10291931
|
DWK Life Sciences
258526104 |
4000 mL | 1 |
6340.00 NOK
Each |
Estimated Shipment: 21-05-2024
Log in to see stock. |
||||
10068090
|
DWK Life Sciences
258526207 |
4000 mL | 2 |
6340.00 NOK
Each |
Estimated Shipment: 21-05-2024
Log in to see stock. |
||||
10787931
|
DWK Life Sciences
258524403 |
1000 mL | 4 |
2500.00 NOK
Each |
Estimated Shipment: 21-05-2024
Log in to see stock. |
||||
10068030
|
DWK Life Sciences
258524103 |
1000 mL | 1 |
1910.00 NOK
Each |
Estimated Shipment: 21-05-2024
Log in to see stock. |
||||
10652941
|
DWK Life Sciences
258524206 |
1000 mL | 2 |
1910.00 NOK
Each |
Estimated Shipment: 21-05-2024
Log in to see stock. |
||||
10653131
|
DWK Life Sciences
258524309 |
1000 mL | 3 |
1910.00 NOK
Each |
Estimated Shipment: 21-05-2024
Log in to see stock. |
||||
10614481
|
DWK Life Sciences
258526404 |
4000 mL | 4 |
7230.00 NOK
Each |
Estimated Shipment: 21-05-2024
Log in to see stock. |
||||
10613711
|
DWK Life Sciences
258524506 |
1000 mL | 5 |
5085.00 NOK
Each |
Estimated Shipment: 21-05-2024
Log in to see stock. |
||||
Description
One of the most important separation methods in the laboratory environment is filtration, which is used to separate or clean different media. For this working method, filters are required which are selected according to the application with regard to size and porosity. Buechner funnels are cylinders with an integrated large-pored sieve plate. Without additional filter paper, only coarse filtration is possible. Buechner funnels can also be used in vacuum filtration. The applied vacuum accelerates the filtration.
- Filter funnels mate to the filtering flask via a conical rubber seal (GUKO)
- From DURAN™ glass with its good thermal shock and chemical resistance
- Fired-on, highly durable white print
Specifications
Boro 3.3 | |
Yes | |
Qualitative inorganic analysis and preparative chemistry; Fine filtration; Fine filtration; Filtration at reduced pressure; Filtering | |
1 Ea. |