Learn More
Lenz Laborglasintrumente Drying Tubes, Bent, with NS Cone
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand: Lenz Laborglasintrumente 05337003
Additional Details : Weight : 0.10000kg
Description
- Drying tubes with NS cone
- Bent (75°)
- Made of DURAN™ tubing
Specifications
NS 24/29 | |
Borosilicate Glass (3.3) |