Learn More
Sampling Systems™ Double-ended Spoon
These double-ended spoons are ideal for taking different sample sizes using the same spoon. The lengths and volume capacities of the spoons vary, allowing for flexibility in sampling both powders and liquids.
86.34 NOK - 172.68 NOK
Specifications
Product Type | Spoon |
---|---|
Material | Stainless Steel |
Autoclavable | Autoclavable |
Disposable | Reusable |
Graduated | No |
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
17962082
|
Sampling Systems™
A104-150 |
150 mm |
86.34 NOK
Each |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17952082
|
Sampling Systems™
A104-180 |
180 mm |
103.63 NOK
Each |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17982072
|
Sampling Systems™
A104-210 |
210 mm |
120.93 NOK
Each |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17952122
|
Sampling Systems™
A104-250 |
250 mm |
143.90 NOK
Each |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17972082
|
Sampling Systems™
A104-300 |
300 mm |
172.68 NOK
Each |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Made from stainless steel
- Ideal for sampling powders and liquids
- Various sizes available
Specifications
Spoon | |
Autoclavable | |
No |
Stainless Steel | |
Reusable | |
Non-sterile |